Assay Details
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||
Literature Links: |
PEX5L PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
| EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
| EUR - Not Available | ||||||||
| AMR - Not Available |
More Information
Set Membership: |
|

PEX5L
Genomic Map