Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMTA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAMTA1 - calmodulin binding transcription activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195563.1 | Intron | NP_001182492.1 | ||||
NM_001242701.1 | Intron | NP_001229630.1 | ||||
NM_015215.3 | Intron | NP_056030.1 | ||||
XM_011541083.2 | Intron | XP_011539385.1 | ||||
XM_011541084.2 | Intron | XP_011539386.1 | ||||
XM_011541085.2 | Intron | XP_011539387.1 | ||||
XM_011541086.2 | Intron | XP_011539388.1 | ||||
XM_011541087.2 | Intron | XP_011539389.1 | ||||
XM_011541088.2 | Intron | XP_011539390.1 | ||||
XM_011541089.2 | Intron | XP_011539391.1 | ||||
XM_011541090.2 | Intron | XP_011539392.1 | ||||
XM_011541091.2 | Intron | XP_011539393.1 | ||||
XM_011541092.2 | Intron | XP_011539394.1 | ||||
XM_017000773.1 | Intron | XP_016856262.1 | ||||
XM_017000774.1 | Intron | XP_016856263.1 | ||||
XM_017000775.1 | Intron | XP_016856264.1 | ||||
XM_017000776.1 | Intron | XP_016856265.1 | ||||
XM_017000777.1 | Intron | XP_016856266.1 | ||||
XM_017000778.1 | Intron | XP_016856267.1 | ||||
XM_017000779.1 | Intron | XP_016856268.1 | ||||
XM_017000780.1 | Intron | XP_016856269.1 | ||||
XM_017000781.1 | Intron | XP_016856270.1 |
LOC105376689 - uncharacterized LOC105376689 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
