Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4921504E06Rik PubMed Links |
| 4921504E06Rik - RIKEN cDNA 4921504E06 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_027600.4 | Intron | NP_081876.2 | ||||

4921504E06Rik
Genomic Map