Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700022H16Rik PubMed Links |
1700022H16Rik - RIKEN cDNA 1700022H16 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Osr1 - odd-skipped related 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
