Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
6430553K19Rik PubMed Links |
6430553K19Rik - RIKEN cDNA 6430553K19 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
