Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2700081O15Rik PubMed Links |
| 2700081O15Rik - RIKEN cDNA 2700081O15 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_175381.6 | 2137 | Silent Mutation | NP_780590.3 | |||
| Mir6991 - microRNA 6991 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Rtn3 - reticulon 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2700081O15Rik
Genomic Map