Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
3100003L05Rik PubMed Links |
| 3100003L05Rik - RIKEN cDNA 3100003L05 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

3100003L05Rik
Genomic Map