Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930412B13Rik PubMed Links |
4930412B13Rik - RIKEN cDNA 4930412B13 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Gm13986 - predicted gene 13986 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
