Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700013G24Rik PubMed Links |
| 1700013G24Rik - RIKEN cDNA 1700013G24 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_027063.2 | 982 | UTR 3 | NP_081339.1 | |||

1700013G24Rik
Genomic Map