Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1500015O10Rik PubMed Links |
1500015O10Rik - RIKEN cDNA 1500015O10 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024283.3 | 719 | Missense Mutation | NP_077245.2 |
Uxs1 - UDP-glucuronate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
