Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1810032O08Rik PubMed Links |
| 1810032O08Rik - RIKEN cDNA 1810032O08 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Gm11744 - predicted gene 11744 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Snord1a - small nucleolar RNA, C/D box 1A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Snord1b - small nucleolar RNA, C/D box 1B | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Snord1c - small nucleolar RNA, C/D box 1C | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| St6galnac2 - ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1810032O08Rik
Genomic Map