Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610100L16Rik PubMed Links |
| 2610100L16Rik - RIKEN cDNA 2610100L16 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Gm30340 - predicted gene, 30340 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Mir124a-2 - microRNA 124a-2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2610100L16Rik
Genomic Map