Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700019B03Rik PubMed Links |
| 1700019B03Rik - RIKEN cDNA 1700019B03 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_029598.1 | 396 | Missense Mutation | NP_083874.1 | |||
| Pex11g - peroxisomal biogenesis factor 11 gamma | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700019B03Rik
Genomic Map