Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700026F02Rik PubMed Links |
1700026F02Rik - RIKEN cDNA 1700026F02 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
