Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700112E06Rik PubMed Links |
| 1700112E06Rik - RIKEN cDNA 1700112E06 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_028275.1 | Intron | NP_082551.1 | ||||

1700112E06Rik
Genomic Map