Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700112J16Rik PubMed Links |
| 1700112J16Rik - RIKEN cDNA 1700112J16 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Lysmd4 - LysM, putative peptidoglycan-binding, domain containing 4 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700112J16Rik
Genomic Map