Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2900026A02Rik PubMed Links |
| 2900026A02Rik - RIKEN cDNA 2900026A02 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_172884.3 | Intron | NP_766472.1 | ||||

2900026A02Rik
Genomic Map