Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700109G14Rik PubMed Links |
1700109G14Rik - RIKEN cDNA 1700109G14 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Arl11 - ADP-ribosylation factor-like 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
