Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
0610040F04Rik PubMed Links |
| 0610040F04Rik - RIKEN cDNA 0610040F04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| 4833447P13Rik - RIKEN cDNA 4833447P13 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

0610040F04Rik
Genomic Map