Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
3830406C13Rik PubMed Links |
| 3830406C13Rik - RIKEN cDNA 3830406C13 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001284383.1 | 407 | Silent Mutation | NP_001271312.1 | |||
| NM_001284384.1 | 407 | Silent Mutation | NP_001271313.1 | |||
| NM_001284385.1 | 407 | Silent Mutation | NP_001271314.1 | |||
| NM_001284386.1 | 407 | Silent Mutation | NP_001271315.1 | |||
| NM_001284387.1 | 407 | Silent Mutation | NP_001271316.1 | |||
| NM_146051.3 | 407 | UTR 3 | NP_666163.1 | |||
| NM_178141.3 | 407 | Silent Mutation | NP_835277.1 | |||

3830406C13Rik
Genomic Map