Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700028P14Rik PubMed Links |
| 1700028P14Rik - RIKEN cDNA 1700028P14 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_026188.2 | Intron | NP_080464.1 | ||||

1700028P14Rik
Genomic Map