Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930425L21Rik PubMed Links |
| 4930425L21Rik - RIKEN cDNA 4930425L21 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

4930425L21Rik
Genomic Map