Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4921530L21Rik PubMed Links |
| 4921530L21Rik - RIKEN cDNA 4921530L21 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_025733.2 | 991 | Missense Mutation | NP_080009.1 | |||

4921530L21Rik
Genomic Map