Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2010016I18Rik PubMed Links |
| 2010016I18Rik - RIKEN cDNA 2010016I18 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Dennd2d - DENN/MADD domain containing 2D | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001093754.2 | Intron | NP_001087223.1 | ||||
| NM_001278941.1 | Intron | NP_001265870.1 | ||||
| NM_028110.2 | Intron | NP_082386.2 | ||||

2010016I18Rik
Genomic Map