Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700017B05Rik PubMed Links |
| 1700017B05Rik - RIKEN cDNA 1700017B05 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_028820.2 | 4194 | UTR 3 | NP_083096.1 | |||
| Trcg1 - taste receptor cell gene 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001014398.2 | 4194 | Intron | NP_001014420.2 | |||

1700017B05Rik
Genomic Map