Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2010111I01Rik PubMed Links |
2010111I01Rik - RIKEN cDNA 2010111I01 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Gm16907 - predicted gene, 16907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
