Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700028E10Rik PubMed Links |
| 1700028E10Rik - RIKEN cDNA 1700028E10 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Stard13 - StAR-related lipid transfer (START) domain containing 13 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001163493.1 | Intron | NP_001156965.1 | ||||
| NM_146258.2 | Intron | NP_666370.3 | ||||

1700028E10Rik
Genomic Map