Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2210018M11Rik PubMed Links |
2210018M11Rik - RIKEN cDNA 2210018M11 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172280.2 | Intron | NP_758484.2 |
