Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700009N14Rik PubMed Links |
| 1700009N14Rik - RIKEN cDNA 1700009N14 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001081095.1 | 408 | Silent Mutation | NP_001074564.1 | |||
| Gm17094 - predicted gene 17094 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700009N14Rik
Genomic Map