Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2010010A06Rik PubMed Links |
| 2010010A06Rik - RIKEN cDNA 2010010A06 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2010010A06Rik
Genomic Map