Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1110008L16Rik PubMed Links |
| 1110008L16Rik - RIKEN cDNA 1110008L16 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_025373.1 | Intron | NP_079649.1 | ||||

1110008L16Rik
Genomic Map