Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700060O08Rik PubMed Links |
| 1700060O08Rik - RIKEN cDNA 1700060O08 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Npas3 - neuronal PAS domain protein 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_013780.2 | 2253 | Silent Mutation | NP_038808.2 | |||

1700060O08Rik
Genomic Map