Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700001K19Rik PubMed Links |
| 1700001K19Rik - RIKEN cDNA 1700001K19 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_025488.2 | Intron | NP_079764.2 | ||||
| Dync1h1 - dynein cytoplasmic 1 heavy chain 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700001K19Rik
Genomic Map