Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Supt3 PubMed Links |
| Supt3 - suppressor of Ty 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_178652.2 | Intron | NP_848767.1 | ||||

Supt3
Genomic Map