Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700016D06Rik PubMed Links |
| 1700016D06Rik - RIKEN cDNA 1700016D06 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024271.1 | Intron | NP_077233.1 | ||||
| Gm7562 - predicted gene 7562 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700016D06Rik
Genomic Map