Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4922502N22Rik PubMed Links |
4922502N22Rik - RIKEN cDNA 4922502N22 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
