Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700030K09Rik PubMed Links |
1700030K09Rik - RIKEN cDNA 1700030K09 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028170.2 | 329 | Missense Mutation | NP_082446.2 |
Calr3 - calreticulin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
