Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700007B14Rik PubMed Links |
| 1700007B14Rik - RIKEN cDNA 1700007B14 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001164235.1 | Intron | NP_001157707.1 | ||||
| NM_027944.1 | Intron | NP_082220.1 | ||||

1700007B14Rik
Genomic Map