Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2810004N23Rik PubMed Links |
| 2810004N23Rik - RIKEN cDNA 2810004N23 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_025615.2 | 463 | Missense Mutation | NP_079891.2 | |||
| Gnpat - glyceronephosphate O-acyltransferase | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

2810004N23Rik
Genomic Map