Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4932441J04Rik PubMed Links |
| 4932441J04Rik - RIKEN cDNA 4932441J04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Pcdh7 - protocadherin 7 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001122758.1 | Intron | NP_001116230.1 | ||||
| NM_018764.2 | Intron | NP_061234.2 | ||||

4932441J04Rik
Genomic Map