Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2310002L09Rik PubMed Links |
| 2310002L09Rik - RIKEN cDNA 2310002L09 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_027104.3 | 874 | Intron | NP_081380.2 | |||
| NM_027981.1 | 874 | UTR 3 | NP_082257.1 | |||

2310002L09Rik
Genomic Map