Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700015E13Rik PubMed Links |
| 1700015E13Rik - RIKEN cDNA 1700015E13 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001039593.1 | 105 | Silent Mutation | NP_001034682.1 | |||
| Gm7694 - predicted gene 7694 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001198955.1 | 105 | Intron | NP_001185884.1 | |||
| Nos1ap - nitric oxide synthase 1 (neuronal) adaptor protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700015E13Rik
Genomic Map