Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700019L22Rik PubMed Links |
| 1700019L22Rik - RIKEN cDNA 1700019L22 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700019L22Rik
Genomic Map