Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610028H24Rik PubMed Links |
| 2610028H24Rik - RIKEN cDNA 2610028H24 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_029816.2 | 3825 | Intron | NP_084092.2 | |||
| Mcm3ap - minichromosome maintenance deficient 3 (S. cerevisiae) associated protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Ybey - ybeY metallopeptidase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_172550.4 | 3825 | UTR 3 | NP_766138.3 | |||

2610028H24Rik
Genomic Map