Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930419G24Rik PubMed Links |
| 4930419G24Rik - RIKEN cDNA 4930419G24 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Pex5l - peroxisomal biogenesis factor 5-like | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

4930419G24Rik
Genomic Map