Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700018B24Rik PubMed Links |
| 1700018B24Rik - enhancer of rudimentary homolog pseudogene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700018B24Rik
Genomic Map