Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2900011O08Rik PubMed Links |
| 2900011O08Rik - RIKEN cDNA 2900011O08 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_144518.3 | Intron | NP_653101.1 | ||||

2900011O08Rik
Genomic Map