Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2510009E07Rik PubMed Links |
| 2510009E07Rik - RIKEN cDNA 2510009E07 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001001881.2 | Intron | NP_001001881.1 | ||||

2510009E07Rik
Genomic Map