Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700080E11Rik PubMed Links |
1700080E11Rik - RIKEN cDNA 1700080E11 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028562.3 | 611 | Missense Mutation | NP_082838.2 |
