Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700007G11Rik PubMed Links |
| 1700007G11Rik - RIKEN cDNA 1700007G11 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001024614.1 | Intron | NP_001019785.1 | ||||

1700007G11Rik
Genomic Map